Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHA3 Mouse anti-Human, Clone: OTI2D1, lyophilized, TrueMAB™
SDP

Catalog No. 501661700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-1700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-1700 Supplier OriGene Supplier No. CF502875
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

PLEKHA3 (Pleckstrin Homology Domain Containing A3) is a Protein Coding gene. Among its related pathways are Metabolism and Cholesterol and Sphingolipids transport / Transport from Golgi and ER to the apical membrane (normal and CF). Gene Ontology (GO) annotations related to this gene include phospholipid binding and 1-phosphatidylinositol binding. An important paralog of this gene is PLEKHA8.

Specifications

Antigen PLEKHA3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2D1
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PLEKHA3
Gene Accession No. Q9HB20
Gene Alias FAPP1
Gene Symbols PLEKHA3
Host Species Mouse
Immunogen Full length human recombit protein of human PLEKHA3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65977
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.