Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156679
Description
PLEKHA4 Polyclonal specifically detects PLEKHA4 in Human samples. It is validated for Western Blot.Specifications
PLEKHA4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PLEKHA4 | |
Synthetic peptides corresponding to PLEKHA4(pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 4) The peptide sequence was selected from the N terminal of PLEKHA4. Peptide sequence LHSGLQMRRARSPDLFTPLSRPPSPLSLPRPRSAPARRPPAPSGDTAPPA The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Bovine: 92%; Canine: 92%; Guinea pig: 92%; Rabbit: 92%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
PEPP-1, PEPP1phosphoinositol 3-phosphate binding protein-1, PH domain-containing family A member 4, phosphoinositol 3-phosphate-binding PH domain protein 1, Phosphoinositol 3-phosphate-binding protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 4, pleckstrin homology domain-containing family A member 4, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 4 | |
Rabbit | |
Affinity purified | |
RUO | |
57664 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction