Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558465 Supplier Invitrogen™ Supplier No. PA558465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSLKRDMEKV ERQAVPQANH TESCHECGRV GPGHTRDCPH RGHDDIVNFE RQEQEGEQYR SQRDPLEGKR DRSKARSPYS PAEEDALFMD LPTGP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.

PLEKHA7 is required for zonula adherens biogenesis and maintenance. Acts via its interaction with KIAA1543/Nezha, which anchors microtubules at their minus-ends to zonula adherens, leading to the recruitment of KIFC3 kinesin to the junctional site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHA7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHA7
Gene Accession No. Q6IQ23
Gene Alias A430081P20Rik; AI414265; DKFZp686M22243; Hadp1; Heart adapter protein 1; PH domain-containing family A member 7; PH domain-containing family A member 7 antibody; pleckstrin homology domain containing A7; pleckstrin homology domain containing A7 antibody; pleckstrin homology domain containing, family A member 2; pleckstrin homology domain containing, family A member 7; pleckstrin homology domain-containing family A member 7; Plekha7
Gene Symbols PLEKHA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHA7. Recombinant protein control fragment (Product #RP-97436).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144100
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.