Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726367 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6367 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6367 Supplier Proteintech Group Inc Supplier No. 105121AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHR1 is a transcription factor known to participate in the regulation of genes involved in phosphate homeostasis and the regulation of genes involved in inter organ sulfate transport. Diseases associated with PHR1 protein mutation include Wallerian Degeneration and deafness due to Autosomal Recessive 63 dysfunction.

Specifications

Antigen PLEKHB1
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PLEKHB1
Gene Accession No. Q9QYE9, Q9UF11, Q9WU68
Gene Alias Evectin 1, EVT1, KPL1, PHR1, PHRET1, PLEKHB1
Gene Symbols Plekhb1
Host Species Rabbit
Immunogen PLEKHB1 Fusion Protein Ag0797
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27276, 58473, 64471
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.