Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PLEKHB2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15683520
![]() |
Novus Biologicals
NBP15683520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156835
![]() |
Novus Biologicals
NBP156835 |
100 μL |
Each for $487.50
|
|
|||||
Description
PLEKHB2 Polyclonal specifically detects PLEKHB2 in Human samples. It is validated for Western Blot.Specifications
PLEKHB2 | |
Polyclonal | |
Rabbit | |
evectin-2, EVT2FLJ20783, FLJ23679, FLJ30436, PH domain-containing family B member 2, pleckstrin homology domain containing, family B (evectins) member 2, pleckstrin homology domain-containing family B member 2 | |
PLEKHB2 | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
55041 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title