Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570205
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 86%; Zebrafish: 75%.
PLEKHB2 gene ontology annotations related to this gene include regulation of cell differentiation.
Specifications
PLEKHB2 | |
Polyclonal | |
Unconjugated | |
PLEKHB2 | |
2310009M15Rik; AU018239; evectin 2; evectin-2; Evt2; evt-2; PH domain-containing family B member 2; Phdc; pleckstrin homology domain containing B2; pleckstrin homology domain containing, family B (evectins) member 2; pleckstrin homology domain containing, family B member 2; pleckstrin homology domain-containing family B member 2; pleckstrin homology domain-containing family B member 2-like; PLEKHB2; si:dkey-121a11.4; similar to pleckstrin homology domain containing, family B (evectins) member 2; zgc:55604 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55041 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96CS7 | |
PLEKHB2 | |
synthetic peptide directed towards the middle region of human PLEKHB2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction