Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584970
Description
Immunogen sequence: YGYGPYGGAY PPGTQVVYAA NGQAYAVPYQ YPYAGLYGQQ PANQVIIRER YRDNDSDLAL GM.
PLEKHB2 gene ontology annotations related to this gene include regulation of cell differentiation.
Specifications
PLEKHB2 | |
Polyclonal | |
Unconjugated | |
PLEKHB2 | |
2310009M15Rik; AU018239; evectin 2; evectin-2; Evt2; evt-2; PH domain-containing family B member 2; Phdc; pleckstrin homology domain containing B2; pleckstrin homology domain containing, family B (evectins) member 2; pleckstrin homology domain containing, family B member 2; pleckstrin homology domain-containing family B member 2; pleckstrin homology domain-containing family B member 2-like; PLEKHB2; si:dkey-121a11.4; similar to pleckstrin homology domain containing, family B (evectins) member 2; zgc:55604 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55041 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CS7 | |
PLEKHB2 | |
Recombinant protein corresponding to Human PLEKHB2. Recombinant protein control fragment (Product #RP-108645). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction