Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120366 Supplier Invitrogen™ Supplier No. PA5120366
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, A-549, BxPC-3, A-431. The target is usually found in the following locations: Peripheral membrane protein, Recycling endosome membrane. Immunogen sequence: MAFVKSGWLL RQSTILKRWK KNWFDLWSDG HLIYYDDQTR QNIEDKVHMP MDCINIRTGQ ECRDTQPPDG KSKDCMLQIV CRDGKTISLC AESTDDCLAW KFTLQDSRTN TAYVGSAVMT DETSVVSSPP PYTAYAAPAP EQAYGYGPYG GAYPPGTQVV YAANGQAYAV PYQYPYAGLY GQQPANQVII RERYRDNDSD LALGMLAGAA TGMALGSLFW VF.

PLEKHB2 gene ontology annotations related to this gene include regulation of cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHB2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PLEKHB2
Gene Accession No. Q96CS7
Gene Alias 2310009M15Rik; AU018239; evectin 2; evectin-2; Evt2; evt-2; PH domain-containing family B member 2; Phdc; pleckstrin homology domain containing B2; pleckstrin homology domain containing, family B (evectins) member 2; pleckstrin homology domain containing, family B member 2; pleckstrin homology domain-containing family B member 2; pleckstrin homology domain-containing family B member 2-like; PLEKHB2; si:dkey-121a11.4; similar to pleckstrin homology domain containing, family B (evectins) member 2; zgc:55604
Gene Symbols PLEKHB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human PLEKHB2 (NP_001094093.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55041
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.