Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732864 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2864 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2864 Supplier Proteintech Group Inc Supplier No. 203891AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.

Specifications

Antigen PLEKHF1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLEKHF1
Gene Accession No. Q3TB82, Q96S99
Gene Alias Apoptosis inducing protein, APPD, LAPF, Phafin 1, PHAFIN1, PLEKHF1, ZFYVE15
Gene Symbols Plekhf1
Host Species Rabbit
Immunogen PLEKHF1 Fusion Protein Ag14228
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72287, 79156
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.