Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531268 Supplier Invitrogen™ Supplier No. PA531268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (82%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PLEKHF2 may play a role in early endosome fusion upstream of RAB5, hence regulating receptor trafficking and fluid-phase transport. Enhances cellular sensitivity to TNF-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PLEKHF2
Gene Accession No. Q9H8W4
Gene Alias 1110070J07Rik; AA673237; EAPF; Endoplasmic reticulum-associated apoptosis-involved protein containing PH and FYVE domains; PH and FYVE domain-containing protein 2; PH domain-containing family F member 2; phafin 2; Phafin2; Phafin-2; pleckstrin homology and FYVE domain containing 2; pleckstrin homology domain containing, family F (with FYVE domain) member 2; pleckstrin homology domain-containing family F member 2; PLEKHF2; ZFYVE18; Zinc finger FYVE domain-containing protein 18
Gene Symbols PLEKHF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PLEKHF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79666
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.