Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524285
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524285 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524285 Supplier Invitrogen™ Supplier No. PA524285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLEKHG1 Polyclonal Antibody for Western Blot, IHC (P)

PLEKHG1 gene ontology annotations related to this gene include regulation of Rho protein signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PLEKHG1
Gene Accession No. Q9ULL1
Gene Alias ARHGEF41; KIAA1209; pleckstrin homology and RhoGEF domain containing G1; pleckstrin homology domain containing, family G (with RhoGef domain) member 1; pleckstrin homology domain-containing family G member 1; PLEKHG1
Gene Symbols PLEKHG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1273-1302 amino acids from the C-terminal region of human PKHG1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.