Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHG2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562523
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2523 Supplier Bethyl Laboratories, Inc Supplier No. A302414A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

PLEKHG2 may be a transforming oncogene with exchange activity for CDC42. PLEKHG2 may be a guanine-nucleotide exchange factor (GEF) for RAC1 and CDC42.PLEKHG2 activated by the binding to subunits beta and gamma of the heterotrimeric guanine nucleotide-binding protein (G protein).

Specifications

Antigen PLEKHG2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PLEKHG2
Gene Accession No. Q9H7P9
Gene Alias Pleckstrin homology domain containing, family G (with RhoGef domain) member 2, CLG, DKFZp667J2325, FLJ00018, FLJ22458, FLJ38638, ARHGEF42
Gene Symbols PLEKHG2
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64857
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.