Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Invitrogen™ PLEKHG3 Polyclonal Antibody
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$617.00
Specifications
| Antigen | PLEKHG3 | 
|---|---|
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | 
| Applications | Immunohistochemistry (Paraffin) | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        PIPA5111713
                    
                    
                        
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Invitrogen™
                         PA5111713  | 
                
            
            
            
            
            
                
                    100 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $617.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        PIPA5111724
                    
                    
                        
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Invitrogen™
                         PA5111724  | 
                
            
            
            
            
            
                
                    100 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $617.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Immunogen sequence: SFESISSLPE VEPDPEAGSE QEVFSAVEGP SAEETPSDTE SPEVLETQLD AHQGLLGMDP PGDMVDFVAA.
PLEKHG3 (Pleckstrin Homology And RhoGEF Domain Containing G3) is a Protein Coding gene. Diseases associated with PLEKHG3 include Congenital Hemolytic Anemia. Gene Ontology (GO) annotations related to this gene include Rho guanyl-nucleotide exchange factor activity. [GeneCards].Specifications
| PLEKHG3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1L390 | |
| 26030 | |
| Recombinant Protein corresonding to Human PLEKHG3. | |
| Antibody | 
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| PLEKHG3 | |
| ARHGEF43; KIAA0599; PH domain-containing family G member 3; pleckstrin homology and RhoGEF domain containing G3; pleckstrin homology domain containing, family G (with RhoGef domain) member 3; pleckstrin homology domain containing, family G, member 3; pleckstrin homology domain-containing family G member 3; PLEKHG3 | |
| PLEKHG3 | |
| Primary | |
| Antigen Affinity Chromatography | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title