Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111713
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111713 100 μL
PIPA5111724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111713 Supplier Invitrogen™ Supplier No. PA5111713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDGGETLYVT ADLTLEDNRR VIVMEKGPLP SPTAGLEESS GQGPSSPVAL LGQVQDFQQS AECQPKEEGS RDPADPSQQG R Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 69%.

PLEKHG3 (Pleckstrin Homology And RhoGEF Domain Containing G3) is a Protein Coding gene. Diseases associated with PLEKHG3 include Congenital Hemolytic Anemia. Gene Ontology (GO) annotations related to this gene include Rho guanyl-nucleotide exchange factor activity. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHG3
Gene Accession No. A1L390
Gene Alias ARHGEF43; KIAA0599; PH domain-containing family G member 3; pleckstrin homology and RhoGEF domain containing G3; pleckstrin homology domain containing, family G (with RhoGef domain) member 3; pleckstrin homology domain containing, family G, member 3; pleckstrin homology domain-containing family G member 3; PLEKHG3
Gene Symbols PLEKHG3
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PLEKHG3 (Product #RP-105353)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.