Learn More
Invitrogen™ PLEKHG4 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PLEKHG4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PLEKHG4 |
| Gene Accession No. | Q58EX7 |
| Gene Alias | 4931414L13Rik; AI448583; ARHGEF44; mFLJ00068; PH domain-containing family G member 4; pleckstrin homology and RhoGEF domain containing G4; pleckstrin homology domain containing, family G (with RhoGef domain) member 4; Pleckstrin homology domain-containing family G member 4; PLEKHG4; PRTPHN1; Puratrophin-1; Purkinje cell atrophy associated protein 1; Purkinje cell atrophy-associated protein 1; RGD1310790; SCA4 |
| Gene Symbols | PLEKHG4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.