Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565968 Supplier Invitrogen™ Supplier No. PA565968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGVGNKAFRDI APSEEAINDR TVNYVLKCRE VRSRASIAVA PFDHDSLYLG ASNSLPGDPA SCSVLGSLNL HLYRDPALLG LRCPLYPSFP Highest antigen sequence identity to the following orthologs - mouse 77%, rat 80%.

Possible role in intracellular signaling and cytoskeleton dynamics at the Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHG4
Gene Accession No. Q58EX7
Gene Alias 4931414L13Rik; AI448583; ARHGEF44; mFLJ00068; PH domain-containing family G member 4; pleckstrin homology and RhoGEF domain containing G4; pleckstrin homology domain containing, family G (with RhoGef domain) member 4; Pleckstrin homology domain-containing family G member 4; PLEKHG4; PRTPHN1; Puratrophin-1; Purkinje cell atrophy associated protein 1; Purkinje cell atrophy-associated protein 1; RGD1310790; SCA4
Gene Symbols PLEKHG4
Host Species Rabbit
Immunogen Recombinant Human PLEKHG4. Recombinant protein control fragment (Product #RP-106657).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.