Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHG4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PLEKHG4B |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PLEKHG4B Polyclonal specifically detects PLEKHG4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PLEKHG4B | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
153478 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:YDRVDEEVHRLVLTSNNRLQQLEHLRELASLLEGNDQQSCQKGLQLAKENPQRTEEMVQDFRRGLSAVVSQAECREGELARWTRSSELCETVS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
pleckstrin homology domain containing, family G (with RhoGef domain) member 4B | |
PLEKHG4B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title