Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHG4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546588 Supplier Invitrogen™ Supplier No. PA546588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HVFLFEDLIL FSKTQKVEGS HDVYLYKQSF KTAEIGMTEN VGDSGLRFEI Sequence homology: Horse: 79%; Human: 100%.

PLEKHG4B (Pleckstrin Homology And RhoGEF Domain Containing G4B) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeRho guanyl-nucleotide exchange factor activity. This gene encodes a large protein that contains a pleckstrin homology domain and may function as a guanine nucleotide exchange factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG4B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLEKHG4B
Gene Accession No. Q96PX9
Gene Alias CTD-2231H16.1; KIAA1909; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family G member 4B; PH domain-containing family G member 4B; pleckstrin homology and RhoGEF domain containing G4B; pleckstrin homology domain containing, family G (with RhoGef domain) member 4B; pleckstrin homology domain-containing family G member 4B; PLEKHG4B
Gene Symbols PLEKHG4B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PLEKHG4B (aa 1021-1070).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153478
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.