Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562010 Supplier Invitrogen™ Supplier No. PA562010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGDRKSTGLK LSKKKARRRH TDDPSKECFT LKFDLNVDIE TEIVPAMKKK SLGEVLLPVF ERKGIALGKV DIYLDQSNTP LSL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Guanine nucleotide exchange factor that activates RHOA and maybe the NF-kappa-B signaling pathway. Involved in the control of neuronal cell differentiation. Plays a role in angiogenesis through regulation of endothelial cells chemotaxis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHG5
Gene Accession No. O94827
Gene Alias BC023181; CMTRIC; DSMA4; GEF720; Guanine nucleotide exchange factor 720; Kiaa0720; Neuronal RhoA GEF protein; NFkB activating protein; novel PH domain-containing protein; PH domain-containing family G member 5; pleckstrin homology and RhoGEF domain containing G5; pleckstrin homology domain containing family G member 5; pleckstrin homology domain containing, family G (with RhoGef domain) member 5; pleckstrin homology domain-containing family G member 5; Plekhg5; RhoA-specific guanine exchange factor SYX1; RhoA-specific guanine exchange factor SYX2; synectin-binding guanine exchange factor; synectin-binding RhoA exchange factor; Syx; Tech; Transcript highly enriched in cortex and hippocampus
Gene Symbols PLEKHG5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHG5. Recombinant protein control fragment (Product #RP-99824).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.