Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHG6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559578
Description
Immunogen sequence: GLLMEVSAET LFGNVPSLIR THRSFWDEVL GPTLEETRAS GQPLDPIGLQ SGFLTFGQRF HPYVQYCLRV KQTMAYAREQ QETNPLFHAF VQWCE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
PLEKHG6 functions as a guanine nucleotide exchange factor activating the small GTPase RHOA, which, in turn, induces myosin filament formation. It activates RHOG but not activate RAC1, or to a much lower extent than RHOA and RHOG. Part of a functional unit, involving MyoGEF, MYH10 and RHOA, at the cleavage furrow to advance furrow ingression during cytokinesis. In epithelial cells, required for the formation of microvilli and membrane ruffles on the apical pole. Along with EZR, required for normal macropinocytosis.
Specifications
PLEKHG6 | |
Polyclonal | |
Unconjugated | |
PLEKHG6 | |
BC026778; Gm155; MyoGEF; myosin II interacting GEF; myosin interacting guanine nucleotide exchange factor; myosin-interacting guanine nucleotide exchange factor; PH domain-containing family G member 6; pleckstrin homology and RhoGEF domain containing G6; pleckstrin homology domain containing, family G (with RhoGef domain) member 6; pleckstrin homology domain-containing family G member 6; PLEKHG6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55200 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3KR16 | |
PLEKHG6 | |
Recombinant protein corresponding to Human PLEKHG6. Recombinant protein control fragment (Product #RP-97628). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction