Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHH1 Monoclonal Antibody (3-E11)
GREENER_CHOICE

Catalog No. PIMA534890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534890 Supplier Invitrogen™ Supplier No. MA534890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: MCF-7, A431, SH-SY5Y, human kidney tissue, mouse kidney tissue.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3-E11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene PLEKHH1
Gene Accession No. Q9ULM0
Gene Alias KIAA1200; PH domain-containing family H member 1; pleckstrin homology domain containing, family H (with MyTH4 domain) member 1; pleckstrin homology domain-containing family H member 1; pleckstrin homology, MyTH4 and FERM domain containing H1; PLEKHH1
Gene Symbols PLEKHH1
Host Species Mouse
Immunogen Recombinant protein within Human PLEKHH1 aa 1,059-1,364.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57475
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.