Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561573
Description
Immunogen sequence: SSSTVHSGET VEAKPLQPHL GRESPPHQPC MKLLTFRCSS ASWGEGLVTA QRGMLPGTKT SAREGGPGSS LTLPKV Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

Specifications
PLEKHH1 | |
Polyclonal | |
Unconjugated | |
PLEKHH1 | |
KIAA1200; PH domain-containing family H member 1; pleckstrin homology domain containing, family H (with MyTH4 domain) member 1; pleckstrin homology domain-containing family H member 1; pleckstrin homology, MyTH4 and FERM domain containing H1; PLEKHH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULM0 | |
PLEKHH1 | |
Recombinant protein corresponding to Human PLEKHH1. Recombinant protein control fragment (Product #RP-100899). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction