Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHH2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306559B
This item is not returnable.
View return policy
Description
PLEKHH2 Polyclonal antibody specifically detects PLEKHH2 in Human samples. It is validated for ImmunofluorescenceSpecifications
PLEKHH2 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
130271 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Biotin | |
KIAA2028, pleckstrin homology domain containing, family H (with MyTH4 domain) member 2, PLEKHH1L | |
Produced in rabbits immunized with E. coli-derived Human PLEKHH2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction