Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543998
Description
Peptide sequence: WQLLALCVGL FLPHHPFLWL LRLHLKRNAD SRTEFGKYAI YCQRCVERTQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
PLEKHH2 contains 1 FERM domain, 1 MyTH4 domain and 2 PH domains. It is a Single-pass membrane protein. The exact function of PLEKHH2 remains unknown.
Specifications
| PLEKHH2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHH2 | |
| AI256725; E030001K05; KIAA2028; mKIAA2028; pleckstrin homology domain containing, family H (with MyTH4 domain) member 2; pleckstrin homology domain-containing family H member 2; pleckstrin homology, MyTH4 and FERM domain containing H2; PLEKHH1L; PLEKHH2; RGD1304935; si:ch73-105b23.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 130271 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IVE3 | |
| PLEKHH2 | |
| Synthetic peptide directed towards the C-terminal of human PLEKHH2 (aa 1043-1092). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction