Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567446 Supplier Invitrogen™ Supplier No. PA567446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STLSSHTSEEG VQCSRMGSEM YLTASDDSSS IFEEETFGIK RPEHKKLYSW QQEAQWKALN SPLGKGNSE Highest antigen sequence identity to the following orthologs - mouse 74%, rat 77%.

PLEKHH2 contains 1 FERM domain, 1 MyTH4 domain and 2 PH domains. It is a Single-pass membrane protein. The exact function of PLEKHH2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHH2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHH2
Gene Accession No. Q8IVE3
Gene Alias AI256725; E030001K05; KIAA2028; mKIAA2028; pleckstrin homology domain containing, family H (with MyTH4 domain) member 2; pleckstrin homology domain-containing family H member 2; pleckstrin homology, MyTH4 and FERM domain containing H2; PLEKHH1L; PLEKHH2; RGD1304935; si:ch73-105b23.3
Gene Symbols PLEKHH2
Host Species Rabbit
Immunogen Recombinant Human PLEKHH2. Recombinant protein control fragment (Product #RP-108108).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.