Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHH3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531398
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (92%), Rat (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLEKHH3, a gene related to endothelial tight junction, is associated with obstructive sleep apnea (OSA) and angiogenesis. It has been reported that the expression of PLEKHH3 protein was increased in the OSA patients. PLEKHH3 may play a key role in OSA and angiogenesis by specifically regulating the endothelial cells formation of tight junctions.
Specifications
PLEKHH3 | |
Polyclonal | |
Unconjugated | |
PLEKHH3 | |
BC020025; PH domain-containing family H member 3; pleckstrin homology domain containing, family H (with MyTH4 domain) member 3; pleckstrin homology domain-containing family H member 3; pleckstrin homology, MyTH4 and FERM domain containing H3; Plekhh3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q7Z736 | |
PLEKHH3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 200 of Human PLEKHH3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction