Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561192 Supplier Invitrogen™ Supplier No. PA561192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYWQLLTCMS CTFRPGGAVQ GHLLGHLERT EQALPDSEPA EYARFIRKAL GRTRGRELVP SLAEISALSQ RQELLCTVHC PGAGACAVAI DS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

PLEKHH3, a gene related to endothelial tight junction, is associated with obstructive sleep apnea (OSA) and angiogenesis. It has been reported that the expression of PLEKHH3 protein was increased in the OSA patients. PLEKHH3 may play a key role in OSA and angiogenesis by specifically regulating the endothelial cells formation of tight junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHH3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHH3
Gene Accession No. Q7Z736
Gene Alias BC020025; PH domain-containing family H member 3; pleckstrin homology domain containing, family H (with MyTH4 domain) member 3; pleckstrin homology domain-containing family H member 3; pleckstrin homology, MyTH4 and FERM domain containing H3; Plekhh3
Gene Symbols PLEKHH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHH3. Recombinant protein control fragment (Product #RP-98342).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.