Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHJ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561857 Supplier Invitrogen™ Supplier No. PA561857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FIEDPERKYH FECSSEEQCQ EWMEALRRAS YEFMRRSLIF YRNEIRKVTG KDPLEQFGIS EEARFQLSGL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 87%.

The PLEKHJ1 (Pleckstrin Homology Domain Containing, Family J Member 1) gene encodes a protein that is involved in signal transduction and cytoskeletal organization. Located on chromosome 11, the PLEKHJ1 protein contains a pleckstrin homology (PH) domain, which facilitates its binding to phosphoinositides and the subsequent interaction with cellular membranes. This interaction is vital for various cellular processes, including the regulation of cell shape, motility, and intracellular trafficking. The precise biological functions of PLEKHJ1 are still being elucidated, but its role in these signaling pathways underscores its importance in maintaining cellular architecture and function. Alterations in PLEKHJ1 expression or mutations can have significant implications for cellular dynamics and have been linked to certain pathological conditions, including cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHJ1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHJ1
Gene Accession No. Q9NW61
Gene Alias 9530063M10Rik; Gnrpx; guanine nucleotide releasing protein x; guanine nucleotide-releasing protein x; PH domain-containing family J member 1; pleckstrin homology domain containing J1; pleckstrin homology domain containing, family J member 1; pleckstrin homology domain-containing family J member 1; Plekhj1
Gene Symbols PLEKHJ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHJ1. Recombinant protein control fragment (Product #RP-98922).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.