Learn More
Invitrogen™ PLEKHJ1 Polyclonal Antibody

Description
Recommended positive controls: HeLa, Jurkat, NT2D1, mouse liver. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PLEKHJ1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PLEKHJ1 |
| Gene Accession No. | Q9D240, Q9NW61 |
| Gene Alias | 9530063M10Rik; Gnrpx; guanine nucleotide releasing protein x; guanine nucleotide-releasing protein x; PH domain-containing family J member 1; pleckstrin homology domain containing J1; pleckstrin homology domain containing, family J member 1; pleckstrin homology domain-containing family J member 1; Plekhj1 |
| Gene Symbols | PLEKHJ1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.