Learn More
Invitrogen™ PLEKHM1 Polyclonal Antibody

Description
A suggested positive control is human lung tissue lysate. PA5-20857 can be used with blocking peptide PEP-0971.
Specifications
Specifications
| Antigen | PLEKHM1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PLEKHM1 |
| Gene Accession No. | Q9Y4G2 |
| Gene Alias | 162 kDa adapter protein; AP162; B2; BC038943; D330036J23Rik; KIAA0356; OPTB6; PH domain-containing family M member 1; pleckstrin homology and RUN domain containing M1; pleckstrin homology domain containing, family M (with RUN domain) member 1; pleckstrin homology domain-containing family M member 1; PLEKHM1 |
| Gene Symbols | PLEKHM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.