Learn More
Invitrogen™ PLEKHM1 Polyclonal Antibody

Description
Immunogen sequence: PARIIHNWDL TKRPICRQAL KFLTQIRAQP LINLQMVNAS LYEHVERMHL IGR Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Specifications
Specifications
| Antigen | PLEKHM1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PLEKHM1 |
| Gene Accession No. | Q9Y4G2 |
| Gene Alias | 162 kDa adapter protein; AP162; B2; BC038943; D330036J23Rik; KIAA0356; OPTB6; PH domain-containing family M member 1; pleckstrin homology and RUN domain containing M1; pleckstrin homology domain containing, family M (with RUN domain) member 1; pleckstrin homology domain-containing family M member 1; PLEKHM1 |
| Gene Symbols | PLEKHM1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.