Learn More
Invitrogen™ PLEKHM3 Polyclonal Antibody

Description
A suggested positive control is mouse skeletal muscle tissue lysate. PA5-20851 can be used with blocking peptide PEP-0965.
Specifications
Specifications
| Antigen | PLEKHM3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PLEKHM3 |
| Gene Accession No. | Q6ZWE6, Q8BM47 |
| Gene Alias | 9430067K14Rik; A230102O09Rik; AI449570; Dapr; differentiation associated protein; Differentiation-associated protein; PH domain-containing family M member 3; pleckstrin homology domain containing M3; pleckstrin homology domain containing, family M, member 1-like; pleckstrin homology domain containing, family M, member 3; pleckstrin homology domain-containing family M member 3; PLEKHM1L; Plekhm3; RGD1307151 |
| Gene Symbols | PLEKHM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.