Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHM3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988245
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8245 Supplier Bioss Supplier No. BS8064R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLEKHM3, also known as DAPR, is a member of the M family of Pleckstrin homology domain-containing proteins. PLEKHM3 was initially identified through chromatin immunoprecipitation and CpG microarray analysis examining proteins regulated by myocyte-enhancing factor 2. In C2C12 myoblast cells, PLEKHM3 binds to the PI3K signaling member protein kinase B in the cytosol prior to differentiation into myotubes. Following the initiation of differentiation, PLEKHM3 was also found in membrane fractions. Knockdown of PLEKHM3 expression by RNAi resulted in the inhibition of myotube formation, suggesting that PLEKHM3 is a key component required by myoblasts for orchestrating their differentiation during myogenesis.

Specifications

Antigen PLEKHM3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLEKHM3
Gene Accession No. Q6ZWE6
Gene Alias 9430067K14Rik; A230102O09Rik; AI449570; DAPR; Differentiation associated protein; Differentiation-associated protein; PH domain-containing family M member 3; pleckstrin homology domain containing M3; pleckstrin homology domain containing, family M, member 1-like; pleckstrin homology domain containing, family M, member 3; Pleckstrin homology domain-containing family M member 3; PLEKHM1L; PLEKHM3; RGD1307151
Gene Symbols PLEKHM3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PLEKHM3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389072
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.