Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556842
Description
Immunogen sequence: IFSEELDGLC FKGELPLRAV HINLEEKEKQ IRSFLIEGPL INTIRVVCAS YEDYGHWLLC LRAVTHREGA PPLPGAESFP GSQV Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.

Specifications
PLEKHN1 | |
Polyclonal | |
Unconjugated | |
PLEKHN1 | |
Cardiolipin and phosphatidic acid-binding protein; CLPABP; PH domain-containing family N member 1; pleckstrin homology domain containing N1; pleckstrin homology domain containing, family N member 1; pleckstrin homology domain-containing family N member 1; PLEKHN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q494U1 | |
PLEKHN1 | |
Recombinant protein corresponding to Human PLEKHN1. Recombinant protein control fragment (Product #RP-94523). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction