Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543108 Supplier Invitrogen™ Supplier No. PA543108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MMKKNNSAKR GPQDGNQQPA PPEKVGWVRK FCGKGIFREI WKNRYVVLKG Sequence homology: Cow: 86%; Human: 100%; Pig: 86%; Rat: 82%.

CKIP1 was identified through a yeast-two hybrid screening as a protein that would interact with only one of the two catalytic subunits of the casein kinase 2 complex. CKIP1 is a pleckstrin homology domain-containing protein localized within the nucleus and at the plasma membrane that interacts with CK2alpha but not CK2alpha', and is thought to play a role in targeting CK2alpha to a particular cellular location. CKIP1 has been implicated in muscle differentiation and the regulation of cell morphology and actin cytoskeleton. CKIP1 can also interact with other proteins such as ATM, an upstream kinase of p53, and recruit the nuclear ATM to the plasma membrane, suggesting CKIP1 may regulate ATM function through re-localizing ATM. CKIP1 can also form a complex with the kinase AKT, leading to a down-regulation of PI3K/AKT signaling and suppression of tumor growth in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLEKHO1
Gene Accession No. Q53GL0
Gene Alias 2810052M02Rik; Casein kinase 2-interacting protein 1; C-Jun-binding protein; CK2 interacting protein 1; CK2 interacting protein 1; HQ0024c protein; CK2-interacting protein 1; CKIP1; CKIP-1; HQ0024c; HQ0024c protein; JBP; Jun zipper-associated 2; Jza2; JZA-20; OC120; Osteoclast maturation-associated gene 120 protein; PH domain-containing family O member 1; pleckstrin homology domain containing O1; pleckstrin homology domain containing, family O member 1; pleckstrin homology domain-containing family O member 1; pleckstrin homology domain-containing family O member 1-like; PLEKHO1; RGD1311487; RP11-458I7.3; TNF intracellular domain-interacting protein
Gene Symbols PLEKHO1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PLEKHO1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51177
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.