Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLEKHO2 Monoclonal Antibody (3D9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524490
Description
Peptide Sequence: GDLLGEGPRH PLQPRERLYR AQLEVKVASE QTEKLLNKVL GSEPAPVSAE TLLSQAVEQL RQATQVLQEM RDLGELSQEA PGLREKRKEL VTLYRRSAP.
Pleckstrin homology domain-containing proteins family O member 2 (PLEKHO2)'s function remains largely unknown. Genetic polymorphisms of PLEKHO2 contribute to diabetic retinopathy (DR) development. Functional prediction analysis strengthened the likelihood of participation of PLEKHO2 in the development of DR (Eric C. Han et al., 2012).
Specifications
| PLEKHO2 | |
| Monoclonal | |
| Unconjugated | |
| PLEKHO2 | |
| AI840980; PH domain-containing family O member 2; PH domain-containing family Q member 1; PH domain-containing protein; PH domain-containing protein homolog; pleckstrin homology domain containing O2; pleckstrin homology domain containing, family O member 2; pleckstrin homology domain containing, family Q member 1; pleckstrin homology domain-containing family O member 2; Pleckstrin homology domain-containing family Q member 1; PLEKHO2; Plekhq1; PP1628; pp9099 | |
| Mouse | |
| Protein A | |
| RUO | |
| 80301 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3D9 | |
| PBS with no preservative; pH 7.4 | |
| Q8TD55 | |
| PLEKHO2 | |
| pp9099 (NP_079477, 392 a.a. ~ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction