Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559046 Supplier Invitrogen™ Supplier No. PA559046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVLPDKLKVS WENPSPQEAP AAESAEPSQA PCSETSEAAP REGGKPPTPP PKILSEKLKA SMGEMQASGP PAPGTVQVSV NGMDD Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 59%.

Pleckstrin homology domain-containing proteins family O member 2 (PLEKHO2)'s function remains largely unknown. Genetic polymorphisms of PLEKHO2 contribute to diabetic retinopathy (DR) development. Functional prediction analysis strengthened the likelihood of participation of PLEKHO2 in the development of DR (Eric C. Han et al., 2012).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHO2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHO2
Gene Accession No. Q8TD55
Gene Alias AI840980; PH domain-containing family O member 2; PH domain-containing family Q member 1; PH domain-containing protein; PH domain-containing protein homolog; pleckstrin homology domain containing O2; pleckstrin homology domain containing, family O member 2; pleckstrin homology domain containing, family Q member 1; pleckstrin homology domain-containing family O member 2; Pleckstrin homology domain-containing family Q member 1; PLEKHO2; Plekhq1; PP1628; pp9099
Gene Symbols PLEKHO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHO2. Recombinant protein control fragment (Product #RP-98130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80301
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.