Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Plet1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572251
Description
Plet1 Polyclonal Antibody for Western Blot
Modulates leading keratinocyte migration and cellular adhesion to matrix proteins during a wound-healing response and promotes wound repair. May play a role during trichilemmal differentiation of the hair follicle.
Specifications
| Plet1 | |
| Polyclonal | |
| Unconjugated | |
| Plet1 | |
| 0610037B23Rik; 1600029D21Rik; AA407923; AgK114; Antigen mAgK114; AU043048; C11orf34; placenta expressed transcript 1; placenta-expressed transcript 1 protein; Plet1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 76509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8VEN2 | |
| Plet1 | |
| KLH conjugated synthetic peptide between 82-115 amino acids from the Central region of Mouse Plet1. | |
| 80 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction