Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin A1 Antibody (708954), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB65361L
Description
Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
Plexin A1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
708954 | |
Flow Cytometry, Immunocytochemistry | |
NOV, NOVP, plexin A1, PLEXIN-A1 | |
E. coli-derived recombinant human Plexin A1, Tyr964-Leu1152, Accession # Q9UIW2 | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience | |
5361 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction