Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plexin-A1 Monoclonal Antibody (7B8H9)
GREENER_CHOICE

Catalog No. PIMA531857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531857 Supplier Invitrogen™ Supplier No. MA531857
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31857 has been tested in indirect ELISA.

Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin-A1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7B8H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PLXNA1
Gene Accession No. Q9UIW2
Gene Alias 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV
Gene Symbols PLXNA1
Host Species Mouse
Immunogen Purified recombinant fragment of human PLXNA1 (AA: 1100-1200) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5361
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.