Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plexin-A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517147 Supplier Invitrogen™ Supplier No. PA517147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin-A1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 50 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PLXNA1
Gene Accession No. Q9UIW2
Gene Alias 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV
Gene Symbols PLXNA1
Host Species Rabbit
Immunogen Synthetic peptide surrounding Gln1875 of human Plexin A1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362398, 5361
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.