Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Plexin-A1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547612
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A2 and rmPlexin A3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
Specifications
Plexin-A1 | |
Polyclonal | |
Unconjugated | |
PLXNA1 | |
2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
Goat | |
Antigen affinity chromatography | |
RUO | |
18844, 5361 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P70206, Q9UIW2 | |
PLXNA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Plexin A1 Ser28-Pro1242. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction