Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plexin-A1 Recombinant Rabbit Monoclonal Antibody (2A5Z6)
GREENER_CHOICE

Catalog No. PIMA538091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538091 Supplier Invitrogen™ Supplier No. MA538091
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: K-562, Mouse brain, Rat testis Immunogen sequence: SVQSCPHGLG STDNVNKLLL LDYAANRLLA CGSASQGICQ FLRLDDLFKL GEPHHRKEHY LSSVQEAGSM AGVLIAGPPG QGQAKLFVGT PIDGKSEYFP T.

Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin-A1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2A5Z6
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene PLXNA1
Gene Accession No. P70206, Q9UIW2
Gene Alias 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV
Gene Symbols PLXNA1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18844, 362398, 5361
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.