Learn More
Invitrogen™ Plexin A2 Monoclonal Antibody (583603)

Description
In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B2, D1, recombinant mouse Plexin A1, A3, B3, or C1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Plexin A2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 583603 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PLXNA2 |
| Gene Accession No. | O75051, P70207 |
| Gene Alias | 2810428A13Rik; AA589422; AW457381; FLJ11751; FLJ30634; KIAA0463; LOW QUALITY PROTEIN: plexin-A2; mKIAA0463; OCT; plex 2; PlexA2; plexin 2; plexin A2; plexin-2; plexin-A2; PLXN2; Plxna2; Semaphorin receptor OCT; si:dkey-164k14.1; transmembrane protein OCT; UNQ209/PRO235 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.