Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin A2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985548
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5548 Supplier Bioss Supplier No. BS3638R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plexins are a family of large, transmembrane receptors for multiple classes of semaphorins in vertebrates. Plexins are widely expressed, and regions of their extracellular domain are homologus to both scatter factor receptors and semaphorin domains. Plexins may act as semaphorin receptors alone or in combination with neuropilins. Plexins are divided into four subfamilies designated plexin-A, -B, -C, and -D. Drosophila plexin-A is a receptor for class I semaphorins and controls motor and axon guidance. Plexin-A3 mediates cell-repelling cues. Plexins B and C are receptors for Sema 4 and Sema 7, respectively.

Specifications

Antigen Plexin A2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLXNA2
Gene Accession No. O75051
Gene Alias 2810428A13Rik; AA589422; AW457381; FLJ11751; FLJ30634; Kiaa0463; mKIAA0463; OCT; Plex 2; PlexA2; plexin 2; plexin A2; plexin-2; Plexin-A2; PLXN2; PLXNA2; Semaphorin receptor OCT; si:dkey-164k14.1; transmembrane protein OCT; UNQ209/PRO235
Gene Symbols PLXNA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Plexin A2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.