Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin B1 Antibody (559830), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB37491D
Description
Plexin B1 Monoclonal antibody specifically detects Plexin B1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Plexin B1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5364 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
559830 | |
Flow Cytometry, CyTOF-ready | |
plexin B1, SEP | |
NS0 mouse myeloma cell line transfected with human Plexin B1, Accession # O43157 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction