Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plexin B1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984906
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4906 Supplier Bioss Supplier No. BS2693R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.

Specifications

Antigen Plexin B1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Plxnb1
Gene Accession No. O43157
Gene Alias 2900002G15Rik; AU042020; KIAA0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; Plexin-B1; PLXN5; PLXNB1; Semaphorin receptor SEP; SEP
Gene Symbols Plxnb1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Plexin B1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5364
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.