Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin B2 Antibody (772417), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6836APCCY7
Description
Plexin B2 Monoclonal antibody specifically detects Plexin B2 in Mouse samples. It is validated for Flow CytometrySpecifications
Plexin B2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
772417 | |
Flow Cytometry | |
MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin B2, Leu20-Trp1029, Accession # B2RXS4 | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers | |
23654 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction