Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin D1 Antibody (752829), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4160PEATT594
Description
Plexin D1 Monoclonal antibody specifically detects Plexin D1 in Human samples. It is validated for Flow CytometrySpecifications
Plexin D1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23129 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
752829 | |
Flow Cytometry | |
KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1 | |
Mouse myeloma cell line NS0-derived recombinant human Plexin D1, Leu47-Ala1271, Accession # Q9Y4D7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction