Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323327
Description
Plexin D1 Polyclonal antibody specifically detects Plexin D1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Plexin D1 | |
Polyclonal | |
Western Blot 1:500 | |
KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1 | |
Plexin D1 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the semaphorin domain from human Plexin D1. This sequence is well conserved in rat and mouse Plexin D1, and has low homology to other Plexin family members. [UniProt# Q9Y4D7] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
23129 | |
Store at -20°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction